Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-1 Mouse anti-Human, Clone: 18D4, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Desmoglein-1
Clone 18D4
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25329020
View Documents
Novus Biologicals
NBP25329020UG
20 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NB253290100
View Documents
Novus Biologicals
NBP253290100UG
100 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NB253290200
View Documents
Novus Biologicals
NBP253290200UG
200 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Desmoglein-1 Monoclonal antibody specifically detects Desmoglein-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, SDS-Page.
Specifications

Specifications

Desmoglein-1
Mouse
Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
18D4
IgG1 Kappa
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit