Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-1 Mouse anti-Human, Clone: 27B2- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$378.35

Specifications

Antigen Desmoglein-1
Clone 27B2
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254531100 View Documents Novus Biologicals
NBP254531100UG
100 ug Each for $378.35
Add to cart
 
NB254531200 View Documents Novus Biologicals
NBP254531200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Desmoglein-1 Monoclonal antibody specifically detects Desmoglein-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications

Specifications

Desmoglein-1
Mouse
Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
27B2
IgG1 Kappa
Human
RUO
Purified
Primary
PBS with No Preservative

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit