Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-1 Mouse anti-Human, Clone: 27B2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253291200UG
Description
Desmoglein-1 Monoclonal antibody specifically detects Desmoglein-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, SDS-Page.Specifications
Desmoglein-1 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1 | |
Mouse | |
Protein A or G purified | |
RUO | |
1828 | |
Store at 4C. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
27B2 | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
DSG1 | |
Human desmoglein-1 intracellular domain (exact sequence is proprietary) | |
200 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction