Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-1 Mouse anti-Human, PE/Atto594, Clone: 18D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254532PEATT594

Catalog No. N254532P594


Add to cart

Description

Description

Desmoglein-1 Monoclonal antibody specifically detects Desmoglein-1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Desmoglein-1
18D4
Flow Cytometry
Conjugated, Purified
DSG1
Human desmoglein-1 intracellular domain (exact sequence is proprietary)
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1
Mouse
IgG1 Kappa
0.1 mL
Primary
1828.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit