Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein 2 Monoclonal Antibody (HL2036), Invitrogen™

Rabbit Monoclonal Antibody

Supplier:  Thermo Scientific MA547263

Encompass

Catalog No. PIMA547263


Add to Cart

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Desmoglein 2 Monoclonal antibody specifically detects Desmoglein 2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Desmoglein 2
Monoclonal
1 mg/mL
PBS with no preservative
O55111, Q14126
DSG2
Recombinant protein encompassing a sequence within the C-terminus region of human Desmoglein 2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL2036
Unconjugated
DSG2
AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Rabbit
Protein A
RUO
13511, 1829
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit