Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-2 Mouse anti-Human, Alexa Fluor™ 350, Clone: 8 E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254529AF350

Catalog No. NP2429A30


Add to cart

Description

Description

Desmoglein-2 Monoclonal antibody specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

Desmoglein-2
8 E5
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
DSG2
Recombinant human full-length DSG2 protein
0.1 mL
Primary
1829
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit