Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-3 Mouse anti-Human, Biotin, Clone: 6G2C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237259B

Catalog No. NBP237259B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Desmoglein-3
6G2C11
Western Blot, ELISA, Immunohistochemistry
Purified
DSG3
Purified recombinant fragment of human DSG3 (AA: 55-159) expressed in E. Coli.
Protein A or G purified
Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction
Primary
1830
Western Blot, ELISA, Immunohistochemistry
Biotin
PBS with 0.05% Sodium Azide
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit