Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-3 Mouse anti-Human, Clone: DSG3/2840, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Desmoglein-3
Clone DSG3/2840
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123258
View Documents
Novus Biologicals
NBP30789220UG
20 μg
Each for $138.37
Add to cart
 
NB123257
View Documents
Novus Biologicals
NBP30789210UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Desmoglein-3
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA
Recombinant fragment (around aa 379-491) human Desmoglein-3 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
DSG3/2840
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction
10 mM PBS with 0.05% BSA
1830
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit