Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Destrin Mouse anti-Human, Canine, Monkey, PE/Atto594, Clone: OTI2F7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271669PEATT594

Catalog No. N27169PE594


Add to cart

Description

Description

Destrin Monoclonal antibody specifically detects Destrin in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Destrin
OTI2F7
Flow Cytometry
Affinity Purified, Conjugated
DSTN
Full length recombinant protein of human Destrin (NP_006861) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
ACTDPdestrin, Actin-depolymerizing factor, ADFbA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), bA462D18.2, destrin (actin depolymerizing factor), DSN
Mouse
IgG1
0.1 mL
Primary
11034.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit