Learn More
Destrin Mouse anti-Canine, Human, Non-human primate, Clone: OTI2F7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525472
Catalog No. PIMA525472
Add to cart
Description
Description
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
Specifications
Destrin
Monoclonal
0.4 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P60981
ACTDP, ADF, bA462D18.2, actin-depolymerizing factor, bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), destrin
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11034, 477147, 695969
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2F7
Unconjugated
DSTN
Liquid
DSTN
Full length human recombit protein of DSTN produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate