Learn More
Microbiologics™ Desulfotomaculum nigrificans derived from ATCC 7946
SDP
Everything needed to grow reference cultures for QC testing is included in one device. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab. Available in a 2 pack, universally used for all types of microbiological control testing.
Supplier:  Microbiologics™ 01026P
Catalog No. 23-047-108
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Biosafety Level: 1
  • 2 self-contained units of a single organism
Specifications
Specifications
ATCC 7946
2°C to 8°C
United States
Microbial Identification
2 KWIK-STIK
1
KWIK-STIK 2 Pack
KWIK-STIK
Foil Sleeves
Safety and Handling
Safety and Handling
ShelfLife : 8 Months (Minimum)