Alfa Aesar™ Desyl bromide, 97%

Manufacturer: Alfa Aesar™ H3269114

Quantity 25g

 View more versions of this product

Catalog No. AAH3269114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1484-50-0
    Molecular Formula C14H11BrO
    Formula Weight 275.14
    MDL Number MFCD00000136
    UN Number UN3261
    Synonym 2-Bromo-2-phenylacetophenone; 2-Bromo-1,2-diphenylethan-1-one
    Chemical Name or Material Desyl bromide

    Specifications

    Formula Weight 275.14
    TSCA TSCA
    Molecular Formula C14H11BrO
    MDL Number MFCD00000136
    CAS 1484-50-0
    UN Number UN3261
    Synonym 2-Bromo-2-phenylacetophenone; 2-Bromo-1,2-diphenylethan-1-one
    Health Hazard 2 GHS H Statement
    H314-H318-H335
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 54° to 56°C
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Quantity 25g
    Health Hazard 1 Danger
    Chemical Name or Material Desyl bromide
    Assay 97%

    GHS H Statement
    H314-H318-H335
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit