Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Desyl chloride, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Quantity | 5g |
Melting Point | 64°C to 69°C |
UN Number | UN3261 |
Beilstein | 744474 |
Formula Weight | 230.69 |
Percent Purity | 98% |
Chemical Name or Material | Desyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 207-181-7
RTECSNumber : AM6825000
TSCA : Yes
Recommended Storage : Ambient temperatures