Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Desyl chloride, 98%

Click to view available options
Quantity:
5g
25g
100g
447-31-4
C14H11ClO
230.691
MFCD00000858
RXDYOLRABMJTEF-UHFFFAOYSA-N
desyl chloride,2-chloro-2-phenylacetophenone,alpha-chlorodeoxybenzoin,1,2-diphenyl-2-chloroethanone,ethanone, 2-chloro-1,2-diphenyl,alpha-chlorobenzyl phenyl ketone,2-chloro-1,2-diphenylethone,acetophenone, alpha-chloro-alpha-phenyl,2-chloro-1,2-diphenyl-ethanone,acetophenone, 2-chloro-2-phenyl
95343
2-chloro-1,2-diphenylethanone
C1=CC=C(C=C1)C(C(=O)C2=CC=CC=C2)Cl
447-31-4
C14H11ClO
230.691
MFCD00000858
RXDYOLRABMJTEF-UHFFFAOYSA-N
desyl chloride,2-chloro-2-phenylacetophenone,alpha-chlorodeoxybenzoin,1,2-diphenyl-2-chloroethanone,ethanone, 2-chloro-1,2-diphenyl,alpha-chlorobenzyl phenyl ketone,2-chloro-1,2-diphenylethone,acetophenone, alpha-chloro-alpha-phenyl,2-chloro-1,2-diphenyl-ethanone,acetophenone, 2-chloro-2-phenyl
95343
2-chloro-1,2-diphenylethanone
C1=CC=C(C=C1)C(C(=O)C2=CC=CC=C2)Cl
Quantity 100g
Melting Point 64°C to 69°C
UN Number UN3261
Beilstein 744474
Formula Weight 230.69
Percent Purity 98%
Chemical Name or Material Desyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 207-181-7

RTECSNumber : AM6825000

TSCA : Yes

Recommended Storage : Ambient temperatures