Learn More
Detecto solo Digital Clinical Scale, Mechanical Height Rod, 550 lb x 0.2 lb / 250 kg x 0.1 kg
Supplier:  Detecto SOLO
Encompass_Preferred
DETECTO’s solo eye-level clinical scale is the most economical, high-quality digital physician scale with height rod available. The solo scale features 550 lb x 0.2 lb / 250 kg x 0.1 kg capacity, 14 in W x 15 in D x 2.4 in H low-profile platform, inline mechanical height rod (measures 30 in to 84.5 in / 76 cm to 214.5 cm), eye-level weight display and column, one-inch-high LCD readouts, Body Mass Index calculation, integral wheels for mobility, one RS232 serial port, and convenient audible beep confirmation. The solo indicator features 8 simple buttons that are easily identifiable in any language.
Catalog No. NC1655553