Learn More
Detergent, Neutral Reagent, Fisher Chemical
Components: Triethylene Glycol (<1.5%), SDS (3.0%), Borates (each <1.0%), EDTA (<2.0%), and Water (to balance)
$154.00 - $438.00

Specifications
Chemical Name or Material | Detergent, Neutral Reagent |
---|---|
CAS | 151-21-3, 6381-92-6, 112-27-6, 1303-96-4 |
MDL Number | MFCD00283948 |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
SF118-20
|
20L | PolyPac™ |
Each for $438.00
Order in Bulk
Request bulk or custom formats |
|
SF118-4
|
4L | Poly Bottle |
Each for $154.00
Case of 4 EA for $491.34
Order in Bulk
Request bulk or custom formats |
|
Specifications
Detergent, Neutral Reagent | |
151-21-3, 6381-92-6, 112-27-6, 1303-96-4 | |
MFCD00283948 |
Safety and Handling

CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.