Learn More
Fisherbrand™ Deuterium Lamps
Deuterium Replacement Lamps are economical, without sacrificing quality
Description
- Economical option without sacrificing product quality
- Manufactured in an ISO-accredited facility
- Full traceability of each lamp from manufacturer through to customer delivery
Specifications
Replacement Lamp | |
Deuterium | |
Applied Biosystems 757, 759, 783 A, 785 A, 1000 S, 980, 120 A, 130 A |