Learn More
ORS Nasco DeWalt™ Cordless Drill/Drivers
Manufacturer:  ORS Nasco 115DC9096
 View more versions of this product
Catalog No. 18-861-227
Add to cart
Description
Description
Compact size allows users to fit into tight spaces
  • Lightweight design
  • Dual speed range 0-500/0-1700rpm delivers optimal performance
  • 1/2 in. (2.54cm) ratcheting chuck reduces bit slippage
  • High-performance frameless motor delivers maximum power of 410 unit watts out (18V model); 300 watts (14.4V model)
  • LED work light provides increased visibility in confined spaces
Warranty and Services
Three years
Specifications
Specifications
Safety
DeWalt drill/driver
Work in tight spaces that requires compact-sized tools
18V
Rechargeable battery pack