Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone Acetate, Anhydrous, Micronized, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
1177-87-3
C24H31FO6
434.50
AKUJBENLRBOFTD-RPRRAYFGSA-N
2-[(1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O
1177-87-3
C24H31FO6
434.50
AKUJBENLRBOFTD-RPRRAYFGSA-N
2-[(1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O

DE122, 1177-87-3, C24H31FO6

Spectrum™ Dexamethasone Acetate, Anhydrous, Micronized, USP is corticosteroid that has strong anti-inflammatory and immunosuppressant effects. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 0.4%
Packaging Poly Pail
Quantity 1 kg
Specific Rotation +82° to +88°
CAS Max % 100%
Percent Purity 97 to 102%