Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone Sodium Phosphate, Powder, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
2392-39-4
C22H28FNa2O8P
516.41
PLCQGRYPOISRTQ-FCJDYXGNSA-L
disodium (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
[Na+].[Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O
2392-39-4
C22H28FNa2O8P
516.41
PLCQGRYPOISRTQ-FCJDYXGNSA-L
disodium (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
[Na+].[Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O

DE123, 2392-39-4, C22H28FNa2O8P

Spectrum™ Dexamethasone Sodium Phosphate, Powder, USP is corticosteroid that has strong anti-inflammatory and immunosuppressant effects. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 1 kg
Specific Rotation +74°-+82°
CAS Max % 100%
Percent Purity 97 to 102%