Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexchlorpheniramine Maleate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2438-32-6
C20H23ClN2O4
390.86
DBAKFASWICGISY-DASCVMRKSA-N
(2Z)-but-2-enedioic acid; [(3S)-3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
OC(=O)\C=C/C(O)=O.CN(C)CC[C@@H](C1=CC=C(Cl)C=C1)C1=CC=CC=N1
2438-32-6
C20H23ClN2O4
390.86
DBAKFASWICGISY-DASCVMRKSA-N
(2Z)-but-2-enedioic acid; [(3S)-3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
OC(=O)\C=C/C(O)=O.CN(C)CC[C@@H](C1=CC=C(Cl)C=C1)C1=CC=CC=N1

DE127, 2438-32-6, C16H19ClN2•C4H4O4

Spectrum™ Dexchlorpheniramine Maleate, USP is an antihistamine with anticholinergic properties that is used to treat allergies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.5%
Ignition Residue 0.2%
Packaging Amber Glass Bottle
Specific Rotation +39.5 to +43.0°
Quantity 100 g
CAS Max % 100%
UN Number UN2811
Percent Purity 98 to 102%