Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dexpanthenol, USP, 98-102%, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
100 g
1 kg
45 kg
Packaging:
Amber Glass Bottle
Poly Drum
Description
Spectrum™ Chemical Dexpanthenol, USP acts on the gastrointestinal tract and increases lower intestinal motility. It is also applied topically to the skin to relieve itching and to promote healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Ignition Residue | 0.1% |
Packaging | Amber Glass Bottle |
Refractive Index | 1.495-1.502 |
Quantity | 1 kg |
Specific Rotation | +29.0° to +31.5° |
Percent Purity | 98 to 102% |
Grade | USP |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction