Learn More
Selleck Chemical LLCSupplier Diversity Partner Dexrazoxane HCl (ICRF-187) 25mg 149003-01-0 ADR-529  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S122225MG
Dexrazoxane HCl (ICRF-187, ADR-529) is an intracellular iron chelator, which decreases the formation of superoxide radicals, used as a cardioprotective agent; also an inhibitor of topoisomerase II
Catalog No. 50-625-0