Learn More
Medchemexpress LLC Dexrazoxane hydrochloride | 149003-01-0 | 99.8% | C11H18Cl2N4O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY762015MG
Encompass_Preferred
Dexrazoxane hydrochloride is a heart protectant used to preserve ovarian function and fertility. It exhibits antioxidant and anti-inflammatory properties and can cross the blood-brain barrier. It improves motor function disorders and offers neuroprotective effects, making it useful in neurodegenerative disease research. This EDTA derivative chelates iron, which reduces metal ion complexing with anthracycline and decreases superoxide radical formation. It protects the heart from cardiotoxic side effects of anthracyclines like doxorubicin.
  • Helps preserve ovarian function and fertility
  • Possesses antioxidant and anti-inflammatory properties
  • Crosses the blood-brain barrier
  • Improves motor function disorders
  • Provides neuroprotective effects for neurodegenerative disease studies
  • Chelates iron, reducing superoxide radical formation
  • Protects the heart from anthracycline cardiotoxicity
Catalog No. 50-004-34511