Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran 150,000, Powder, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9004-54-0
9004-54-0
CAS Max % 1
Appearance White to Slightly Off White
Grade Reagent
Loss on Drying 0.1
Packaging Fiber Drum
pH 4.0 to 7.0
Quantity 25 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.