Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dextran, 500,000, Powder, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
100 g
1 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Description
Spectrum™ Chemical Dextran, 500,000, Powder is a complex branched glucan that is synthethized by certain lactic acid bacteria from sucrose. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.
Specifications
Color | White powder |
CAS Max % | 100% |
Ash | 2% |
Quantity | 1 kg |
Grade | Reagent |
Loss on Drying | 10% |
Packaging | Poly Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction