Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran, 500,000, Powder, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
9004-54-0
C18H32O16
504.44
FZWBNHMXJMCXLU-UHFFFAOYNA-N
2,3,4,5-tetrahydroxy-6-{[3,4,5-trihydroxy-6-({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}hexanal
OCC1OC(OCC2OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C2O)C(O)C(O)C1O
9004-54-0
C18H32O16
504.44
FZWBNHMXJMCXLU-UHFFFAOYNA-N
2,3,4,5-tetrahydroxy-6-{[3,4,5-trihydroxy-6-({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}hexanal
OCC1OC(OCC2OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C2O)C(O)C(O)C1O

D1004, 9004-54-0

Spectrum™ Dextran, 500,000, Powder is a complex branched glucan that is synthethized by certain lactic acid bacteria from sucrose. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.
CAS Max % 1
Appearance White powder
Ash 0.02
Loss on Drying 0.1
Packaging Fiber Drum
Quantity 50 kg