Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextran sulfate sodium salt, approx. M.W. 500,000, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
9011-18-1
C9H11NO2
165.192
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine,ethyl aminobenzoate,ethyl p-aminobenzoate,americaine,anesthesin,anaesthesin,ethoform,norcaine,orthesin,parathesin
2337
CHEBI:116735
ethyl 4-aminobenzoate
CCOC(=O)C1=CC=C(C=C1)N
9011-18-1
C9H11NO2
165.192
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine,ethyl aminobenzoate,ethyl p-aminobenzoate,americaine,anesthesin,anaesthesin,ethoform,norcaine,orthesin,parathesin
2337
CHEBI:116735
ethyl 4-aminobenzoate
CCOC(=O)C1=CC=C(C=C1)N
Merck Index 15,2953
Solubility Information Solubility in water: 100g/L (25°C)
Avg. Mol. Wt. or Mol. Wt. Range approx. 50000g/mol
Physical Form Powder
Chloride 0.01% max.
Infrared Spectrum Authentic
Iron (Fe) 0.1% max.
Loss on Drying 10% max.
Packaging Glass bottle
Solubility colorless to light yellow
Sulfated Ash 40% to 50% ()
Color White to Yellow
Quantity 5g
Chemical Name or Material Dextran sulfate sodium salt
Show More Show Less