Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextromethorphan Hydrobromide, Monohydrate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
6700-34-1
C18H28BrNO2
370.33
STTADZBLEUMJRG-IKNOHUQMSA-N
hydrogen (1S,9S,10S)-4-methoxy-17-methyl-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2,4,6-triene hydrate bromide
[H+].O.[Br-].COC1=CC=C2C[C@H]3[C@H]4CCCC[C@@]4(CCN3C)C2=C1
6700-34-1
C18H28BrNO2
370.33
STTADZBLEUMJRG-IKNOHUQMSA-N
hydrogen (1S,9S,10S)-4-methoxy-17-methyl-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2,4,6-triene hydrate bromide
[H+].O.[Br-].COC1=CC=C2C[C@H]3[C@H]4CCCC[C@@]4(CCN3C)C2=C1

DE139, 6700-34-1, C18H25NO•HBr•H2O

Spectrum™ Dextromethorphan Hydrobromide, Monohydrate, USP is as a cough suppressant, for the temporary relief of cough. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Ignition Residue 0.001
Quantity 12 kg
CAS Max % 1
UN Number UN3077
Percent Purity 98 to 102%