Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose, Anhydrous, Granular, BiotechGrade, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
50-99-7
C6H12O6
180.16
WQZGKKKJIJFFOK-UHFFFAOYNA-N
6-(hydroxymethyl)oxane-2,3,4,5-tetrol
OCC1OC(O)C(O)C(O)C1O
50-99-7
C6H12O6
180.16
WQZGKKKJIJFFOK-UHFFFAOYNA-N
6-(hydroxymethyl)oxane-2,3,4,5-tetrol
OCC1OC(O)C(O)C(O)C1O

D1082, 50-99-7, C6H12O6

Spectrum™ Dextrose, Anhydrous, Granular, BiotechGrade is a simple monosaccharide sugar and is commonly referred to as glucose. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Packaging Poly Bottle
Quantity 2.5 kg
Appearance White, granular powder
CAS Max % 1