Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  J.T.Baker 191601

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Catalog No. 02-002-708

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

50-99-7
180.16
Baker Analyzed™ ACS Reagent
C6H12O6
Dextrose, Anhydrous

Specifications

1L = 1.54kg
50-99-7
C6H12O6
Poly Bottle
Baker Analyzed™ ACS Reagent
500g
Powder
180.16
Dextrose, Anhydrous
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit