Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dextrose, Monohydrate, Powder, USP, 97.5-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
77938-63-7
C6H14O7
198.17
SPFMQWBKVUQXJV-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal hydrate
O.OCC(O)C(O)C(O)C(O)C=O
77938-63-7
C6H14O7
198.17
SPFMQWBKVUQXJV-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal hydrate
O.OCC(O)C(O)C(O)C(O)C=O

DE145, 77938-63-7, C6H12O6•H2O

Spectrum™ Dextrose, Monohydrate, Powder, USP - All Spectrum Chemical USP products, including all USP dextrose products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Quantity 45 kg
CAS Max % 1
Percent Purity 97.5 to 102%