Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFF40/CAD, Rabbit anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP176884AF700

Catalog No. NB004464


Add to cart

Description

Description

DFF40/CAD Polyclonal specifically detects DFF40/CAD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

DFF40/CAD
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.-
Rabbit
IgG
0.1 mL
Apoptosis
Primary
Human
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
DFFB
Antibody was raised against a 16 amino acid peptide near the amino terminus of human DFF40. The immunogen is located within the first 50 amino acids of DFF40.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit