Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DFF45/ICAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$241.00 - $518.50
Specifications
Antigen | DFF45/ICAD |
---|---|
Applications | Western Blot, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DFF45/ICAD Polyclonal specifically detects DFF45/ICAD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
DFF45/ICAD | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DFF1DNA fragmentation factor 45 kDa subunit, DFF45DNA fragmentation factor subunit alpha, DFF-45DNA fragmentation factor, 45 kD, alpha polypeptide, DNA fragmentation factor, 45kDa, alpha polypeptide, ICAD, Inhibitor of CAD | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human DFF45/ICAD (NP_004392.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Tumor Suppressors | |
PBS with 50% glycerol, pH7.3. | |
1676 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title