Learn More
Tocris Bioscience™ DFHBI Available on GSA/VA Contract for Federal Government customers only.
Mimic of GFP fluorophore for imaging RNA in living cells
$188.70 - $782.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
560910 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5609/10
Each for $188.70
Add to cart
560950 View Documents Tocris Bioscience™
5609/50
Each for $782.00
Add to cart
Description
Description
Mimic of green fluorescent protein (GFP) fluorophore for imaging RNA in living cells. Fluorescence is activated by binding to Spinach2 or Broccoli aptamers. Exhibits peak excitation maxima of 447 nm and peak fluorescence emission of 501 nm when bound to Spinach2.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.