Learn More
SHOWA™ DFlex™ G4 Cut Resistant Gloves
Cut resistant glove offers enhanced wet grip
Manufacturer:  SHOWA™ 8210D2S10
 View more versions of this product
Catalog No. 19-168-930
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • G4 composite yarn construction
  • Smooth grip
  • Ambidextrous
  • Fully launderable
  • Color-coded cuff indicates size
  • Thickness: 10 gauge
  • Color: blue
Specifications
Specifications
Granular
Knit
mechanical and automotive, construction, machine or compartment assembly, recycling, bottling, Glass handling, sheet Metal handling
10
Level 4
Ambidextrous
HPPE (High-performance Polyethylene), PE (Polyethylene)
Blue