Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFNA5 Mouse anti-Human, Purified MaxPab™ Polyclonal , Abnova™

Mouse polyclonal antibody raised against a full-length human DFNA5 protein.

Manufacturer:  Abnova Corporation H00001687B02P

Catalog No. 89-934-042


Add to cart

Description

Description

Hearing impairment is a heterogeneous condition with over 40 loci described. The protein encoded by this gene is expressed in fetal cochlea, however, its function is not known. Nonsyndromic hearing impairment is associated with a mutation in this gene. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MFAKATRNFLREVDADGDLIAVSNLNDSDKLQLLSLVTKKKRFWCWQRPKYQFLSLTLGDVLIEDQFPSPVVVESDFVKYEGKFANHVSGTLETALGKVKLNLGGSSRVESQSSFGTLRKQEVDLQQLIRDSAERTINLRNPVLQQVLEGRNEVLCVLTQKITTMQKCVISEHMQVEEKCGGIVGIQTKTVQVSATEDGNVTKDSNVVLEIPAATTIAYGVIELYVKLDGQFEFCLLRGKQGGFENKKRIDSVYLDPLVFREFAFIDMPDAAHGISSQDGPLSVLKQATLLLERNFHPFAELPEPQQTALSDIFQAVLFDDELLMVLEPVCDDLVSGLSPTVAVLGELKPRQQQDLVAFLQLVGCSLQGGCPGPEDAGSKQLFMTAYFLVSALAEMPDSAAALLGTCCKLQIIPTLCHLLRALSDDGVSDLEDPTLTPLKDTERFGIVQRLFASADISLERLKSSVKAVILKDSKVFPLLLCITLNGLCALGREHS
Specifications

Specifications

DFNA5
Polyclonal
Mouse polyclonal antibody raised against a full-length human DFNA5 protein.
DFNA5
Purified
DFNA5
DFNA5 (NP_004394.1,1 a.a. ∽ 496 a.a) full-length human protein.
Protein A/G purification
RUO
Antibody
Polyclonal
Antibody reactive against mammalian transfected lysate.
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_004403
ICERE-1
Mouse
IgG
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1687
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit