Learn More
Tocris Bioscience™ DG 172 dihydrochloride Available on GSA/VA Contract for Federal Government customers only.
Potent PPARβ/δ inverse agonist; promotes BMC differentiation
$186.60 - $775.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
556650 View Documents Tocris Bioscience™
5566/50
Each for $775.00
Add to cart
556610 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5566/10
Each for $186.60
Add to cart
Description
Description
Potent PPARβ/δ inverse agonist (IC50 = 26.9 nM). Inhibits Angptl4 gene expression in mouse myoblasts (IC50 = 9.5 nM) in vitro. Also augments GM-CSF/IL-4-induced differentiation of mature dendritic cells from bone marrow cells in culture. Orally available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.