Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DGCR14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551543
Description
Immunogen sequence: LRVEGSETPY VDRTPGPAFK ILEPGRRERL GLKMANEAAA KNRAKKQEAL RRVTENLASL TPKGLSPAMS PALQRLVSRT ASKYTDRALR ASYTPSPARS THLKTPASGL QTPTSTPAPG SATRTPLTQD PASITDNL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
May be involved in pre-mRNA splicing.
Specifications
DGCR14 | |
Polyclonal | |
Unconjugated | |
ESS2 | |
AI462402; D16H22S1269E; Dgcr1; DGCR13; DGCR14; DGSH; DGS-H; DGSI; DGS-I; DiGeorge syndrome critical region 13; DiGeorge syndrome critical region 14; diGeorge syndrome critical region 14 homolog; DiGeorge syndrome critical region gene 13; DiGeorge syndrome critical region gene 14; DiGeorge syndrome critical region gene 14 homolog; DiGeorge syndrome critical region gene DGSI; DiGeorge syndrome critical region protein 14; DiGeorge syndrome gene H; DiGeorge syndrome gene I; diGeorge syndrome protein H; ES2; ES2 protein; Es2el; ESS2; Expressed sequence 2 embryonic lethal; Protein DGCR13; protein DGCR14; Protein ES2; Splicing factor ESS-2 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27886, 360741, 8220 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O70279, Q96DF8 | |
DGCR14, ESS2 | |
Recombinant protein corresponding to Human DGCR14. Recombinant protein control fragment (Product #RP-91371). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction