Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGCR14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549435

Catalog No. PA549435

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in pre-mRNA splicing.
Specifications

Specifications

DGCR14
Polyclonal
PBS with 0.09% sodium azide
Q96DF8, O70279
Protein DGCR14, DiGeorge syndrome critical region 13, DiGeorge syndrome critical region 14, DiGeorge syndrome protein H, DGS-H, Protein ES2, DGCR14, DGCR13, DGSH, DGSI, ES2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27886, 360741, 8220
Western Blot
Unconjugated
DGCR14
Liquid
DGCR14
KLH conjugated synthetic peptide between 365-398 amino acids from the C-terminal region of human DGCR14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit