Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGCR2 Mouse anti-Human, Mouse, Rat, Clone: 1019908, R&D Systems™
Mouse Monoclonal Antibody
$136.67 - $385.56
Specifications
Antigen | DGCR2 |
---|---|
Clone | 1019908 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10443100
|
R&D Systems
MAB10443100 |
100 μg |
Each for $385.56
|
|
MAB10443SP
|
R&D Systems
MAB10443SP |
25 μg |
Each for $136.67
|
|
Description
DGCR2 Monoclonal specifically detects DGCR2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DGCR2 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DGS-C, DiGeorge syndrome critical region gene 2, IDDDKFZp686I1730, integral membrane protein deleted in DiGeorge syndrome, integral membrane protein DGCR2/IDD, KIAA0163, KIAA0163DiGeorge syndrome critical region protein 2, LAN, SEZ-12 | |
Chinese Hamster Ovary cell line CHO-derived human DGCR2, Glu22-Ala349, Accession # P98153 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1019908 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P98153 | |
9993 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title