Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGK-alpha Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI4A11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270547JF549

Catalog No. N270547J549


Add to cart

Description

Description

DGK-alpha Monoclonal antibody specifically detects DGK-alpha in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

DGK-alpha
OTI4A11
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
DGKA
Full length recombinant protein of human DGK-alpha (NP_001336) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
1606.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit