Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGK-beta Antibody (OTI1F9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270552AF405
Description
DGK-beta Monoclonal specifically detects DGK-beta in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot.Specifications
DGK-beta | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DGKB | |
Full length recombinant protein of human DGK-beta (NP_663733) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Primate | |
IgG1 |
Western Blot | |
OTI1F9 | |
Western Blot | |
90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
1607.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction