Learn More
Invitrogen™ DGKB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109630
Description
Immunogen sequence: LMKILKDIEN STEIMLDRWK FEVIPNDKDE KGDPVPYSII NNYFSIGVDA SIAHRFHIMR EKHPEKFNSR MKNKFWYFEF GTSETFSATC KKLHESVEIE CDGVQIDLIN ISLEGIAILN IPSMHGGSNL WGESKKRRSH RRIEKKGSDK RTTVTDAKEL KFASQDLSDQ LLEVVGLEGA MEMGQIYTGL KSAGRRLAQC SCVVIRTSKS LPMQIDGEPW MQTPCTVSTE.
Diacylglycerol (DAG) is an allosteric activator of protein kinase C. DAG also participates in regulating RAS and RHO family proteins by activating the guanine nucleotide exchange factors VAV and RASGRP1. DAG is also involved in the synthesis of phospholipids and triacylglycerols. Tight regulation of DAG levels is achieved via DAG kinases (DGKs), which remove DAG by phosphorylate it to phosphatidic acid. Several mammalian isozymes of DAGK have been identified.
Specifications
DGKB | |
Polyclonal | |
Unconjugated | |
DGKB | |
6430574F24; 90 kDa diacylglycerol kinase; 90kda; C630029D13Rik; DAG kinase beta; Dagk; DAGK2; DGK; Dgkb; DGK-beta; Diacylglycerol kinase 90kDa; diacylglycerol kinase beta; diacylglycerol kinase beta, 90kDa; diacylglycerol kinase, beta; diacylglycerol kinase, beta 90kDa; diglyceride kinase beta; Kiaa0718; mKIAA0718 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
217480, 54248 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.2 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P49621, Q6NS52 | |
DGKB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 544-773 of human DGKB (NP_663733.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.