Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHCR7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115116
Description
Antibody detects endogenous levels of total DHCR7.
Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
Specifications
DHCR7 | |
Polyclonal | |
Unconjugated | |
DHCR7 | |
7-dehydrocholesterol reductase; 7-DHC reductase; AA409147; D7SR; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; Dhcr7; putative sterol reductase SR-2; SLOS; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13360, 1717, 64191 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O88455, Q9UBM7, Q9Z2Z8 | |
DHCR7 | |
A synthesized peptide derived from human DHCR7(Accession Q9UBM7), corresponding to amino acid residues M1-P51. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction