Learn More
Drg International Inc DRG DHEA ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA3415
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of dehydroepiandrosterone (DHEA) in serum and plasma. Dehydroepiandrosterone (DHEA; androstenolone; 3b-hydroxy-5-androsten-17-one) is a C19 steroid produced in the adrenal cortex and, to a lesser extent, gonads. DHEA serves as a precursor in testosterone and estrogen synthesis. DHEA has a rapid metabolic clearance rate as compared to its sulfated conjugate, DHEA-S. A variety of in vitro effects, including anti-proliferative effects in different cell lines and effects on enzyme-mediated cell metabolism have been reported. In vivo studies suggest that DHEA may affect cholesterol and lipid metabolism, insulin sensitivity and secretion and immune function. Measurement of serum DHEA is a useful marker of adrenal androgen synthesis. The DRG DHEA ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding.
Catalog No. 50-187-1237
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.