Learn More
Crystal Chem IncSupplier Diversity Partner Dehydroepiandrosterone (DHEA) ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80959
The Dehydroepiandrosterone (DHEA) ELISA is a highly sensitive assay that is used to quantify levels of human DHEA in serum. Highlights: Kit uses sample size of only 50 µL. High sensitivity of 0.1 ng/mL. Dynamic range of 0.2 - 40 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7865