Learn More
Crystal Chem IncSupplier Diversity Partner Dehydroepiandrosterone Sulfate (DHEAS) ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80960
The Dehydroepiandrosterone Sulfate (DHEAS) ELISA is a highly sensitive assay that is used to quantify levels of human DHEAS in serum. Highlights: Kit uses sample size of only 50 µL. High sensitivity of 0.005 µg/mL. Wide dynamic range of 0.005 - 10 µg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7866