Learn More
DHPS Antibody (OTI2C9), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270570V
Catalog No. NBP270570V
Add to Cart
Description
Description
DHPS Monoclonal specifically detects DHPS in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
DHPS
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DHPS
Full length recombinant protein of human DHPS (NP_001921) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot
OTI2C9
Western Blot
deoxyhypusine synthase, DHS, DS, EC 2.5.1, EC 2.5.1.46, hypusine, MIG13, migration-inducing gene 13
Mouse
Affinity Purified
Cell Cycle and Replication
1725.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions