Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHRS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | DHRS1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156415
|
Novus Biologicals
NBP156415 |
100 μL |
Each of 1 for $436.00
|
|
Description
DHRS1 Polyclonal specifically detects DHRS1 in Human samples. It is validated for Western Blot.Specifications
DHRS1 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
Q96LJ7 | |
115817 | |
Synthetic peptides corresponding to DHRS1(dehydrogenase/reductase (SDR family) member 1) The peptide sequence was selected from the N terminal of DHRS1. Peptide sequence ITGRHLDTLRVVAQEAQSLGGQCVPVVCDSSQESEVRSLFEQVDREQQGR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dehydrogenase/reductase (SDR family) member 1, dehydrogenase/reductase SDR family member 1, DKFZp586I0523, EC 1.1, FLJ14250, FLJ25430, MGC20204, SDR19C1, short chain dehydrogenase/reductase family 19C, member 1 | |
DHRS1 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title